Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Cyanide, Technical, Approx. 85%, Spectrum™ Chemical
SDP

Catalog No. 18614746 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-746 500 g Fiber Drum
18-614-744 25 g Fiber Drum
18-614-743 2.5 kg Fiber Drum
18-614-742 12 kg Fiber Drum
18-614-745 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-614-746 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z1066500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 557-21-1

Specifications

CAS Max % 1
Physical Form Powder, Crystals and/or Chunks
Quantity 500 g
Molecular Weight (g/mol) 117.42
Percent Purity ∼85%
Grade Technical
Packaging Fiber Drum
Chemical Name or Material Zinc Cyanide

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.