Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc Diethyldithiocarbamate

Catalog No. 32970325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-970-325G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 32-970-325G Supplier Sigma Aldrich Supplier No. 32970325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinc Diethyldithiocarbamate

Specifications

CAS 87198-17-2
Molecular Formula C3O9Tm2 -+ xH2O
Linear Formula Tm2(CO3)3 -+ xH2O
Quantity 1 g
MDL Number MFCD00149979
Synonym Zinc bis(diethyldithiocarbamate); Zinc diethylcarbamodithioate; Diethyldithiocarbamic acid zinc salt
Molecular Weight (g/mol) 517.9 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.