Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Dimethyldithiocarbamate, MP Biomedicals

Catalog No. ICN210386 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210386 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210386 Supplier MP Biomedicals, Inc Supplier No. 0521038680

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 137-30-4
Molecular Formula C6H12N2S4Zn
Molecular Weight (g/mol) 305.796
InChI Key DUBNHZYBDBBJHD-UHFFFAOYSA-L
Synonym ziram, zinc dimethyldithiocarbamate, carbazinc, fuclasin, methazate, zarlate, aazira, cymate, tsimat, zinc bis dimethyldithiocarbamate
PubChem CID 8722
ChEBI CHEBI:79736
IUPAC Name zinc;N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)[S-].CN(C)C(=S)[S-].[Zn+2]

Specifications

Quantity 100 g
Formula Weight 305.82
Chemical Name or Material Zinc Dimethyldithiocarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.