Learn More
Strem, An Ascensus Company CAS# 7440-66-6. 5000G. Zinc dust (97.5%). MFCD00011291
SDP
Catalog No. 501963543
Encompass
CAS# 7440-66-6. 5000G. Zinc dust (97.5%). MFCD00011291. Molecular Formula: Zn. Molecular Weight: 65.38. Color/Form: -325 mesh. Stability: moisture sensitive. Strem# 93-3056. www.strem.com/catalog/v3/93-3056/
This item is not returnable. View return policy
Catalog No. 50-196-3543 Supplier Strem, An Ascensus Company Supplier No. 9330565000G
CAS# 7440-66-6. 5000G. Zinc dust (97.5%). MFCD00011291. Molecular Formula: Zn. Molecular Weight: 65.38. Color/Form: -325 mesh. Stability: moisture sensitive. Strem# 93-3056. www.strem.com/catalog/v3/93-3056/