Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc ethylene-1,2-bisdithiocarbamate | 12122-67-7 | 275.74 g/mol | C4H6N2S4Zn | 250 MG
SDP

Catalog No. 5000282690
Encompass_Preferred

Zineb is an agricultural fungicide of the dithiocarbamate class supplied for research use. It is a zinc bis(dithiocarbamate) compound with broad-spectrum activity against fungal pathogens, provided as a solid with supplier storage guidance and laboratory pack sizes.

  • Agricultural fungicide of the dithiocarbamate class.
  • Broad-spectrum activity against fungal pathogens.
  • Zinc-containing bis(dithiocarbamate) compound.
  • Solid form, suitable for laboratory handling.
  • CAS number 12122-67-7; formula C4H6N2S4Zn.
  • Molecular weight 275.74 g/mol.
  • Recommended sealed, dry storage; solvent storage conditions noted by supplier.
  • Available in small pack sizes for research, including 250 MG.

Catalog No. 50-002-82690 Supplier Medchemexpress LLC Supplier No. HYW127739250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zineb is an agricultural fungicide of the dithiocarbamate class supplied for research use. It is a zinc bis(dithiocarbamate) compound with broad-spectrum activity against fungal pathogens, provided as a solid with supplier storage guidance and laboratory pack sizes.

  • Agricultural fungicide of the dithiocarbamate class.
  • Broad-spectrum activity against fungal pathogens.
  • Zinc-containing bis(dithiocarbamate) compound.
  • Solid form, suitable for laboratory handling.
  • CAS number 12122-67-7; formula C4H6N2S4Zn.
  • Molecular weight 275.74 g/mol.
  • Recommended sealed, dry storage; solvent storage conditions noted by supplier.
  • Available in small pack sizes for research, including 250 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.