Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Ethylenebisdithiocarbamate, MP Biomedicals

Catalog No. ICN219919 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219919 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219919 Supplier MP Biomedicals, Inc Supplier No. 0521991980

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12122-67-7
Molecular Formula C4H6N2S4Zn
Molecular Weight (g/mol) 275.726
InChI Key AMHNZOICSMBGDH-UHFFFAOYSA-L
Synonym zineb, metiram, parzate, zinc ethylenebisthiocarbamate, zinc ethylenebisdithiocarbamate, cynkotox, novozir, zinc ethane-1,2-diylbis dithiocarbamate, dithane z-78, polikarbatsin
PubChem CID 5284484
ChEBI CHEBI:52498
IUPAC Name zinc;N-[2-(sulfidocarbothioylamino)ethyl]carbamodithioate
SMILES C(CNC(=S)[S-])NC(=S)[S-].[Zn+2]

Specifications

Quantity 100 g
Formula Weight 277.7
Chemical Name or Material Zinc Ethylenebisdithiocarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.