Learn More
Novus Biologicals™ Zinc finger protein 774 Recombinant Protein Antigen

Description
Specifications
Specifications
| Gene ID (Entrez) | 342132 |
| Purification Method | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | Zinc finger protein 774 Recombinant Protein Antigen |
| Content And Storage | Store at −20°C. Avoid freeze-thaw cycles |
| Formulation | PBS and 1M Urea, pH 7.4 |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Alias | MGC75360, zinc finger protein 774 |
| Gene Symbol | ZNF774 |
| Label Type | Unlabeled |
| Product Type | Recombinant Protein Antigen |
| Show More |
For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.