Learn More
Strem, An Ascensus Company CAS# 7783-49-5. 25g. Zinc fluoride, anhydrous, 99%. MFCD00011298
SDP
Supplier:  Strem, An Ascensus Company 93301125G
Encompass
CAS# 7783-49-5. 25g. Zinc fluoride, anhydrous, 99%. MFCD00011298. Molecular Weight: 103.37. Molecular Formula: ZnF2. Color/form: white xtl. Strem# 93-3011. http://www.strem.com/catalog/v/93-3011/
Catalog No. 50-901-14961