Learn More
Strem, An Ascensus Company CAS# 7440-66-6. 1pc. Zinc foil (99.9+%). MFCD00011291
SDP
Catalog No. 5090111348
Encompass
CAS# 7440-66-6. 1pc. Zinc foil (99.9+%). MFCD00011291. Molecular Weight: 65.38. Molecular Formula: Zn. Color/form: 1.6mm thick, 15cm x 15cm (~225g/pc) Strem# 30-0040. http://www.strem.com/catalog/v/30-0040/
This item is not returnable. View return policy
Catalog No. 50-901-11348 Supplier Strem, An Ascensus Company Supplier No. 3000401PC
CAS# 7440-66-6. 1pc. Zinc foil (99.9+%). MFCD00011291. Molecular Weight: 65.38. Molecular Formula: Zn. Color/form: 1.6mm thick, 15cm x 15cm (~225g/pc) Strem# 30-0040. http://www.strem.com/catalog/v/30-0040/