Learn More
Sigma Aldrich Fine Chemicals Biosciences Zinc foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 99.95+% | 7440-66-6 | MFCD00011291 | 10EA
Catalog No. 501880737
Encompass
Zinc foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 99.95+% | Purity: 99.95+ % | Mol Wt: 65.39 | 7440-66-6 | MFCD00011291 | 10EA
This item is not returnable. View return policy
Catalog No. 50-188-0737 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF7773963210EA
Zinc foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 99.95+% | Purity: 99.95+ % | Mol Wt: 65.39 | 7440-66-6 | MFCD00011291 | 10EA