Learn More
Zinc Formalin Fixative, Sigma-Aldrich
Catalog No. 01258879 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3.75 L
Catalog No. Quantity
01-258-879 3.75 L
1 options
Catalog No. 01-258-879 Supplier MilliporeSigma Supelco Supplier No. Z29023.75L
Add to Cart
Add to Cart
Zinc Formalin Fixative is an excellent morphological preservation of nuclear and cytoplasmic components. Can replace neutral buffered formalin for routine tissue and immunohistochemical procedures. It is a non-precipitating fixative which can be used with automated and manual methods.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fixative
Quantity 3.75 L
Solution Type Fixative
Form Liquid