Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc gluconate | 4468-02-4 | MFCD00868110 | 98.0% | 455.67 | C12H22O14Zn | 1 ML
SDP

Catalog No. 5000283081
Encompass_Preferred

Zinc gluconate is the gluconate salt of zinc used as a research reagent in studies of inflammation, immune function, and nutritional supplementation. It is water- and DMSO-soluble, and manufacturer documentation reports a molecular weight of 455.67 and a typical purity of 98.0%.

  • Used in research of inflammation, zinc deficiency, and immune responses.
  • Soluble in DMSO and water at up to 50 mg/mL with ultrasonic assistance.
  • Molecular weight 455.67.
  • Purity 98.0% as supplied.
  • Storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • In vivo formulation guidance provided for achieving ≥2.5 mg/mL in mixed solvent systems.

Catalog No. 50-002-83081 Supplier Medchemexpress LLC Supplier No. HYW14549910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinc gluconate is the gluconate salt of zinc used as a research reagent in studies of inflammation, immune function, and nutritional supplementation. It is water- and DMSO-soluble, and manufacturer documentation reports a molecular weight of 455.67 and a typical purity of 98.0%.

  • Used in research of inflammation, zinc deficiency, and immune responses.
  • Soluble in DMSO and water at up to 50 mg/mL with ultrasonic assistance.
  • Molecular weight 455.67.
  • Purity 98.0% as supplied.
  • Storage: sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • In vivo formulation guidance provided for achieving ≥2.5 mg/mL in mixed solvent systems.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.