Learn More
Medchemexpress LLC Zinc gluconate | 4468-02-4 | MFCD00868110 | 98.0% | 455.67 | C12H22O14Zn | 25 G
SDP
Catalog No. 5000282904
Encompass_Preferred
Zinc gluconate is the zinc salt of gluconic acid provided as a white powder for research and analytical use. It serves as a source of bioavailable zinc ions for biochemical, pharmacological, and nutritional studies, and is supplied with supporting documentation for laboratory handling.
  • High purity (98.0%) for research-grade applications.
  • Anhydrous molecular weight 455.67 and formula C12H22O14Zn.
  • Soluble in DMSO and water at 50 mg/mL with sonication.
  • Supplied as a dry powder; store sealed at 4°C to protect from moisture.
  • Datasheet and certificate of analysis available for lot-specific data.
  • Suitable for biochemical, pharmacological, and nutritional research.
Catalog No. 50-002-82904 Supplier Medchemexpress LLC Supplier No. HYW14549925G
Zinc gluconate is the zinc salt of gluconic acid provided as a white powder for research and analytical use. It serves as a source of bioavailable zinc ions for biochemical, pharmacological, and nutritional studies, and is supplied with supporting documentation for laboratory handling.
  • High purity (98.0%) for research-grade applications.
  • Anhydrous molecular weight 455.67 and formula C12H22O14Zn.
  • Soluble in DMSO and water at 50 mg/mL with sonication.
  • Supplied as a dry powder; store sealed at 4°C to protect from moisture.
  • Datasheet and certificate of analysis available for lot-specific data.
  • Suitable for biochemical, pharmacological, and nutritional research.