Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Gluconate, Hydrate, Spectrum™ Chemical
SDP

Catalog No. 18621708 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-708 50 kg
18-621-705 500 g
18-621-706 2.5 kg
18-621-707 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-621-708 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ZI10850KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4468-02-4

Specifications

Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 98 to 102%
Grade Not Available
Packaging Poly Bottle
Chemical Name or Material Zinc Gluconate, Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.