Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Gluconate, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18607851 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-851 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-851 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z10772.5KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z1077, 4468-02-4, C12H22O14Zn

Spectrum™ Chemical Zinc Gluconate, USP is used for treatment and prevention of zinc deficiency. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Zinc Gluconate, USP, Z1077, C12H22O14Zn

Chemical Identifiers

CAS 4468-02-4
Molecular Formula C12H22O14Zn
Molecular Weight (g/mol) 455.67
MDL Number MFCD00868110
InChI Key WHMDKBIGKVEYHS-IYEMJOQQSA-L
IUPAC Name zinc(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
SMILES [Zn++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

CAS Max % 1
pH 5.5 to 7.5
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Zinc Gluconate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.