Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma Zinc ICP Standard

Catalog No. 1703890100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-038-90100 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-038-90100 Supplier MilliporeSigma Supplier No. 1703890100
Add to Cart
Add to Cart

Certified reference material produced in accordance with ISO 17034, characterized in accordance with ISO/IEC 17025, and traceable to SI unit kg and measured against primary material from a National Metrology Institute.

This certified reference material is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. It's traceable to SI unit kg and measured against primary material from a National Metrology Institute.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc ICP Standard
Packaging Plastic Bottle
Solution Type Calibration Standard
Type Certified Reference Material
Industry Type Industrial QC, Pharmaceutical
Quantity 100 mL
Grade Certified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.