Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc ICP Standard, Ricca Chemical
SDP

Catalog No. 95014 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
9501-4 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 9501-4 Supplier Ricca Chemical Company Supplier No. 95014

1mL = 1mg Zn, 1000ppm Zn.(Zn in 5% HNO3) (Class: 8, UN3264, Pkg Grp: III, Corrosive Liquid, Acidic, Inorganic, n.o.s, Contains: Nitric Acid)

Chemical Identifiers

CAS 7732-18-5

Specifications

Quantity 120 mL
Chemical Name or Material Zinc ICP Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.