Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc(II) ethylenediaminetetraacetic acid disodium tetrahydrate | 176736-49-5 | MFCD00151728 | 95.0% | 471.63 g/mol | C10H20N2Na2O12Zn | 1 KG
SDP

Catalog No. 5000282628
Encompass_Preferred

Zinc(II) ethylenediaminetetraacetic acid disodium tetrahydrate is the zinc salt of EDTA supplied as a crystalline reagent for laboratory and life-science research. It is used as a chelating agent in metal-binding studies, analytical assays, and biochemical applications where controlled zinc complexation is required.

  • Acts as a chelating agent for zinc and other metal ions.
  • Suitable for biochemical assays and analytical applications.
  • Provided as a white to off-white crystalline powder.
  • Purity approximately 95.0% as supplied.
  • Molecular weight 471.63 g/mol; formula C10H20N2Na2O12Zn.
  • Available in a 1 kg package for bulk laboratory use.

Catalog No. 50-002-82628 Supplier Medchemexpress LLC Supplier No. HYW1277761KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinc(II) ethylenediaminetetraacetic acid disodium tetrahydrate is the zinc salt of EDTA supplied as a crystalline reagent for laboratory and life-science research. It is used as a chelating agent in metal-binding studies, analytical assays, and biochemical applications where controlled zinc complexation is required.

  • Acts as a chelating agent for zinc and other metal ions.
  • Suitable for biochemical assays and analytical applications.
  • Provided as a white to off-white crystalline powder.
  • Purity approximately 95.0% as supplied.
  • Molecular weight 471.63 g/mol; formula C10H20N2Na2O12Zn.
  • Available in a 1 kg package for bulk laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.