Learn More
Medchemexpress LLC Zinc(II)-protoporphyrin IX | 15442-64-5 | 98.9% | 626.02 | C34H32N4O4Zn | 5 MG
SDP
Catalog No. 5000212472
Encompass_Preferred
Zinc Protoporphyrin (Zn(II)-protoporphyrin IX) is a porphyrin-based small molecule used in biomedical research as a competitive heme oxygenase-1 (HO-1) inhibitor and as a biochemical marker of iron deficiency. It is supplied as a solid for laboratory studies and should be stored sealed and protected from moisture at low temperatures for stability.
  • Competitive heme oxygenase-1 (HO-1) inhibitor
  • Used as a screening marker of iron deficiency in biological samples
  • High purity suitable for research assays (98.9%)
  • Available in milligram-scale packages for laboratory use
  • Requires sealed, moisture-free storage at low temperature
Catalog No. 50-002-12472 Supplier Medchemexpress LLC Supplier No. HY1011935MG
Zinc Protoporphyrin (Zn(II)-protoporphyrin IX) is a porphyrin-based small molecule used in biomedical research as a competitive heme oxygenase-1 (HO-1) inhibitor and as a biochemical marker of iron deficiency. It is supplied as a solid for laboratory studies and should be stored sealed and protected from moisture at low temperatures for stability.
  • Competitive heme oxygenase-1 (HO-1) inhibitor
  • Used as a screening marker of iron deficiency in biological samples
  • High purity suitable for research assays (98.9%)
  • Available in milligram-scale packages for laboratory use
  • Requires sealed, moisture-free storage at low temperature