Learn More
Medchemexpress LLC Zinc(II)-protoporphyrin IX | 15442-64-5 | MFCD00011612 | 98.9% | 626.02 g/mol | C34H32N4O4Zn | 100 MG
SDP
Catalog No. 5000213369
Encompass_Preferred
Zinc(II)-protoporphyrin IX is a porphyrin derivative used as a heme oxygenase-1 (HO-1) inhibitor in biochemical and pharmacological research. It is applied in studies of oxidative stress, heme metabolism, and related signaling pathways, provided as a stable solid with recommended sealed storage at -20°C for long-term stability.
  • Used as an HO-1 inhibitor for mechanistic and pathway studies.
  • Well-characterized chemical identity with CAS 15442-64-5.
  • High purity suitable for in vitro biochemical assays and preclinical work.
  • Soluble in DMSO; manufacturer provides solubility and vehicle preparation guidance.
  • Stable when stored sealed at -20°C; solvent storage recommendations available.
Catalog No. 50-002-13369 Supplier Medchemexpress LLC Supplier No. HY101193100MG
Zinc(II)-protoporphyrin IX is a porphyrin derivative used as a heme oxygenase-1 (HO-1) inhibitor in biochemical and pharmacological research. It is applied in studies of oxidative stress, heme metabolism, and related signaling pathways, provided as a stable solid with recommended sealed storage at -20°C for long-term stability.
  • Used as an HO-1 inhibitor for mechanistic and pathway studies.
  • Well-characterized chemical identity with CAS 15442-64-5.
  • High purity suitable for in vitro biochemical assays and preclinical work.
  • Soluble in DMSO; manufacturer provides solubility and vehicle preparation guidance.
  • Stable when stored sealed at -20°C; solvent storage recommendations available.