Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc iodide, 99.999%, (trace metal basis), extra pure, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
10139-47-6
I2Zn
319.19
MFCD00011299
10139-47-6
I2Zn
319.19
MFCD00011299
For ultra trace analysis
Assay Percent Range 99.999%
Linear Formula ZnI2
Fieser 01,1294; 05,765; 10,462; 11,604; 12,576; 13,88
Merck Index 15, 10339
Solubility Information Solubility in water: 4500g/L (20°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether
Formula Weight 319.19
Percent Purity 100.00%
Grade Trace Metal Basis
Density 4.7400g/mL
Packaging Glass Bottle
Specific Gravity 4.74
Total Trace Metal Impurities 0.001% max
Melting Point 446°C
Quantity 5g
Chemical Name or Material Zinc iodide, 99.999%
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 233-396-0

TSCA : TSCA