Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc iodide, ultra dry, 99.995% (metals basis)

Catalog No. AA3572706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3572706 5 g
AA3572714 25 g
AA3572722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3572706 Supplier Thermo Scientific Chemicals Supplier No. 03572706
Add to Cart

CAS: 10139-47-6 | ZnI2

Zinc iodide is used as an x-ray opaque penetrant in industrial radiography to enhance the contrast. It is used as electrode compartments and as a stain in electron microscopy in association with osmium tetroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10139-47-6
Molecular Formula ZnI2
MDL Number MFCD00011299

Specifications

Boiling Point 625°C (decomposition)
Melting Point 446°C
Physical Form Granules
Quantity 5 g
Assay Percent Range 99.9%, metal basis
Solubility Information Soluble in water.
Formula Weight 319.18
Sensitivity Hygroscopic
Density 4.74
Chemical Name or Material Zinc Iodide, Ampouled under argon
missing translation for 'healthHazard1' H314
missing translation for 'healthHazard2' Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
missing translation for 'healthHazard3' P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: II
EINECSNumber 233-396-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.