Learn More
Strem, An Ascensus Company CAS# 14074-80-7. 250mg. Zinc meso-tetraphenylporphine. MFCD00012155
SDP
Catalog No. 5081501
Encompass
CAS# 14074-80-7. 250mg. Zinc meso-tetraphenylporphine. MFCD00012155. Molecular Weight: 678.10. Molecular Formula: (C44H28N4)Zn. Color/form: purple xtl. Strem# 30-3500. http://www.strem.com/catalog/v/30-3500/
Catalog No. 50-815-01 Supplier Strem, An Ascensus Company Supplier No. 303500250MG
CAS# 14074-80-7. 250mg. Zinc meso-tetraphenylporphine. MFCD00012155. Molecular Weight: 678.10. Molecular Formula: (C44H28N4)Zn. Color/form: purple xtl. Strem# 30-3500. http://www.strem.com/catalog/v/30-3500/