Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Zinc, Metal Powder
SCI_ED

Catalog No. s25637 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Reagent Grade - Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25637 500 g Reagent Grade - Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25637
Only null left
Add to Cart
Edge
Add to Cart

Reacts with water or moisture

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 6-6-7440
Synonym Zinc Dust

Specifications

Density 7.14g/mL
Quantity 500 g
Packaging Reagent Grade - Solid
Form Solid
Assay Percent Range 100% w/w
Linear Formula Zn
Formula Weight 65.37
Chemical Name or Material Zinc, Metal Powder
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.