Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Naphthenate (Zn ca. 10%), TCI America™
SDP

Catalog No. N034125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N034125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N034125G Supplier TCI America Supplier No. N034125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12001-85-3
MDL Number MFCD00147699
Synonym Naphthenic Acid Zinc Salt

Specifications

Color Brown
Quantity 25 g
UN Number 3082
Chemical Name or Material Zinc Naphthenate (Zn ca. 10%)
EINECSNumber (8)-0630
RTECSNumber QK9275000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.