Learn More
Strem, An Ascensus Company CAS# 10196-18-6. 500g. Zinc nitrate hexahydrate, min. 98%. MFCD00149889
SDP
Supplier:  Strem, An Ascensus Company 933015500G
Encompass
CAS# 10196-18-6. 500g. Zinc nitrate hexahydrate, min. 98%. MFCD00149889. Molecular Weight: 189.41 (297.47). Molecular Formula: Zn(NO3)2∙6H2O. Color/form: white xtl. Strem# 93-3015. http://www.strem.com/catalog/v/93-3015/
Catalog No. 50-901-14968