Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc nitride, 99% (metals basis)

Catalog No. AA8821209
Click to view available options
Quantity:
2 g
10 g

CAS: 1313-49-1 | N2Zn3 | 224.15 g/mol

Zinc nitride is useful in optical and electronic devices. It is doped with oxygen to prepare a potential nitride-based transparent conductor, which is used in semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-49-1
Molecular Formula N2Zn3
Molecular Weight (g/mol) 224.15
MDL Number MFCD00061327
InChI Key AKJVMGQSGCSQBU-UHFFFAOYSA-N
Synonym zinc nitride, zinc nitride zn3n2
PubChem CID 12130759
SMILES [Zn++].[N-]=[Zn].[N-]=[Zn]

Specifications

Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water. Soluble in hydrochloric acid.
IUPAC Name zinc;azanidylidenezinc
Formula Weight 224.12
Percent Purity 99%
Density 6.22 g/mL
Assay (metals basis)
Chemical Name or Material Zinc nitride
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 215-207-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.