Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc oxalate dihydrate, Puratronic™, 99.999% (metals basis)

Catalog No. p-7049130
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1298514 25 g
AA1298522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1298514 Supplier Thermo Scientific Chemicals Supplier No. 01298514
Only null left
Add to Cart

CAS: 55906-21-3 | C2H6O6Zn | 191.44 g/mol

Zinc oxalate dihydrate is used as an chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55906-21-3
MDL Number MFCD00150611

Specifications

Color White
Physical Form Powder
Quantity 25 g
Molecular Formula C2H6O6Zn
Linear Formula ZnC2O4·2H2O
UN Number UN3288
Merck Index 14,10146
Synonym zinc oxalate dihydrate, zinc oxalate, c2o4zn.2h2o
Solubility Information Soluble in dilute mineral acids.
InChI Key PLYRAVNBPDIEOH-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O.O.O.[Zn]
IUPAC Name oxalic acid;zinc;dihydrate
Molecular Weight (g/mol) 191.44
PubChem CID 91886451
Formula Weight 189.42 (153.39 Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Density 3.28 g/mL
Assay (metals basis)
Chemical Name or Material Zinc oxalate dihydrate
Show More Show Less
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 208-934-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.