Learn More
U.S. Pharmacopeia Zinc oxide, 1314-13-2, MFCD00011300, 200 mg
Catalog No. NC1385626
Encompass
Linear Formula: ZnO, Molecular Weight: 81.39, Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis.
Catalog No. NC1385626 Supplier U.S. Pharmacopeia Supplier No. 1724747
Linear Formula: ZnO, Molecular Weight: 81.39, Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis.