Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc oxide

Catalog No. 54490650G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-490-650G 50 g
54-490-610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 54-490-650G Supplier Sigma Aldrich Supplier No. 54490650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinc oxide (ZnO) nanopowder is a metal oxide that is an n-type direct band semiconducting material with a band gap of 3.37 eV. Its properties include high chemical stability, photostability, and high electrochemical coupling coefficient. It is classified as a semiconductor in group II-VI with bond energy of 60 meV, and high thermo-mechanical stability at ambient temperatures.ZnO nanopowder can be used for a variety of applications, such as catalysts, optical materials, UV absorbers, gas sensors and cosmetics.

Specifications

CAS 1314-13-2
Quantity 50 g
Molecular Formula OZn
Linear Formula ZnO
MDL Number MFCD00011300
Molecular Weight (g/mol) 81.39
Percent Purity ∼80%Zn basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.