Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc oxide, dispersion

Catalog No. 721077100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-107-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-107-7100G Supplier Sigma Aldrich Supplier No. 721077100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinc oxide, dispersion (ZnO) is a multi-function metal oxide with unique physio-chemical properties, which includes high chemical stability, broad range absorption spectra and high electrochemical characteristics. It can also be categorized as a semiconductor due to its high energy band width and high thermal stability that make it potentially useful in electronics and optoelectronics applications.ZnO dispersions have piezo and pyroelectric properties that make it useful in the development of sensors, generators, and photo-catalysis. It has low toxicity and biocompatibility, which facilitate its applications in biomedicine.

Specifications

CAS 1314-13-2
Quantity 100 g
Linear Formula ZnO
MDL Number MFCD00011300
Density 1.7 g/mL ±0.1 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.