Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc oxide nanoparticle ink

Catalog No. 7933615ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
50 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-336-15ML 5 mL
79-336-125ML 25 mL
79-336-150ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 79-336-15ML Supplier Sigma Aldrich Supplier No. 7933615ML
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinc oxide nanoparticle ink are nanoparticle-based printing inks allowing processing temperatures of 80°C. Zinc oxide nanoparticle ink is an electron selective interface layer ink containing zinc oxide (ZnO) nanoparticles in isopropanol. ZnO nanoparticle ink functions well as electron extraction layer (EEL) materials in solar cells.This ZnO nanoparticle ink is for slot-dye, spin-coating, and doctor blading for the use as electron transport layer in printed electronics. This ZnO nanoparticle ink is universally applicable in normal and inverted architecture.Annealing temperature <100°C.Prior to application: Ultrasonicate and (optionally) filter through 0.45 mum PTFE filterPost-treatment: Annealing of deposited ZnO films at 80°C - 120CProduct of Avantama Ltd.

Specifications

Density 0.800 g/mL (at 25°C)
Linear Formula ZnO
Quantity 5 mL
Synonym Avantama N-10; Nanograde N-10; Zinc oxide suspension; ZnO dispersion; ZnO ink; ZnO nanoparticle ink
Biological Activity Central nervous system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.