Learn More
MOLECULAR DEPOT LLC Zinc Oxide Nanowires -50 mg
Catalog No. 5000013548
Encompass_Preferred
Zinc oxide Nanowires consists of highly pure zinc oxide nanowires This product has been used in UV lasers solar energy and piezoelectrics devices It has also been used in a wide array of other biochemical applications For in vitro research use only
Catalog No. 50-000-13548 Supplier MOLECULAR DEPOT LLC Supplier No. B2010732
Zinc oxide Nanowires consists of highly pure zinc oxide nanowires This product has been used in UV lasers solar energy and piezoelectrics devices It has also been used in a wide array of other biochemical applications For in vitro research use only