Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Oxide USP Powder, Macron Fine Chemicals™

Catalog No. MK8824212 Shop All Avantor Products
Change view
Click to view available options
Quantity:
2.5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-8824-212 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MK-8824-212 Supplier Avantor Supplier No. 882405
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1314-13-2
Physical Form Powder
Quantity 2.5 kg
Molecular Formula OZn
MDL Number MFCD00011300
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILES O=[Zn]
IUPAC Name oxozinc
Molecular Weight (g/mol) 81.38
PubChem CID 14806
ChEBI CHEBI:36560
Formula Weight 81.39
Grade USP
Packaging STAKMOR
Chemical Name or Material Zinc Oxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.