Learn More
Medchemexpress LLC Zinc phthalocyanine | 14320-04-8 | 98.0% | 577.90 | 25 G
SDP
Catalog No. 5000385462
Encompass_Preferred
Zinc phthalocyanine (ZnPc) is commonly applied in industry as catalysts and photoconductors, and in biomedical fields such as photodynamic therapy (PDT). It can be used to photooxidise cyclohexane and shows promise for solar-cell applications.
  • Commonly applied in industry (catalysts, photoconductors)
  • Applied in biomedical fields (photodynamic therapy, PDT)
  • Can be used to photooxidise cyclohexane
  • Promising for research of solar-cell applications
  • Inhibits the growth of cancer cells Kyse-140 and OE-33
  • Acts as a photosensitizer that produces ROS under specific light irradiation, impairs mitochondrial membrane integrity, activates caspase-3, and induces apoptosis
Catalog No. 50-003-85462 Supplier Medchemexpress LLC Supplier No. 25GHY19204
Zinc phthalocyanine (ZnPc) is commonly applied in industry as catalysts and photoconductors, and in biomedical fields such as photodynamic therapy (PDT). It can be used to photooxidise cyclohexane and shows promise for solar-cell applications.
  • Commonly applied in industry (catalysts, photoconductors)
  • Applied in biomedical fields (photodynamic therapy, PDT)
  • Can be used to photooxidise cyclohexane
  • Promising for research of solar-cell applications
  • Inhibits the growth of cancer cells Kyse-140 and OE-33
  • Acts as a photosensitizer that produces ROS under specific light irradiation, impairs mitochondrial membrane integrity, activates caspase-3, and induces apoptosis