Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc phthalocyanine

Catalog No. 3411695G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-116-95G 5 g
34-116-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 34-116-95G Supplier Sigma Aldrich Supplier No. 3411695G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinc (II) phthalocyanine (ZnPc) complexes show photosensitizing properties. Phthalocyanines show significant +ß+ß interactions and easy formation of H aggregates.Zinc phthalocyanine finds general applications in industry (catalysts, photoconductors etc) and biomedical (photodynamic therapy, PDT). ZnPc may be used to photooxidise cyclohexane. A study reports functionalization of polymer membrane with ZnPc.

Specifications

CAS 14320-04-8
Molecular Formula C32H16N8Zn
Linear Formula C32H16N8Zn
Quantity 5 g
MDL Number MFCD00041955
Synonym Phthalocyanine zinc salt
Molecular Weight (g/mol) 577.91
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.