Learn More
MSC Worksmart S-Hook, Low Carbon Steel, Electrogalvanized, 26 Lb. Capacity, 1 ea.
Catalog No. 5000513593
Encompass
This low carbon steel S-hook provides a reliable hardware solution for marine and industrial applications. It features an electrogalvanized finish for enhanced corrosion resistance.
  • Opening size of 0.219 in. (7/32 in.)
  • Load capacity of 26 lb.
  • Overall length of 1.5 in.
  • Wire size of 0.148 in.
  • Constructed from low-carbon steel
  • Electrogalvanized finish
Catalog No. 50-005-13593 Supplier MSC Supplier No. 13574843
This low carbon steel S-hook provides a reliable hardware solution for marine and industrial applications. It features an electrogalvanized finish for enhanced corrosion resistance.
  • Opening size of 0.219 in. (7/32 in.)
  • Load capacity of 26 lb.
  • Overall length of 1.5 in.
  • Wire size of 0.148 in.
  • Constructed from low-carbon steel
  • Electrogalvanized finish