Learn More
MSC WorkSmart S-Hook, 1/8 in. Opening, Low-Carbon Steel, Electrogalvanized, 1 ea.
Catalog No. 5000513595
Encompass
This low-carbon steel S-hook is designed for versatile use in marine and industrial environments. Featuring an electrogalvanized finish for enhanced corrosion resistance, this hardware is engineered for reliable performance in various rigging and fastening applications.
  • Opening size of 1/8 in.
  • Load capacity of 7 lb.
  • Durable low-carbon steel construction
  • Electrogalvanized zinc coating for added protection
  • Suitable for both industrial and marine applications
Catalog No. 50-005-13595 Supplier MSC Supplier No. 13574876
This low-carbon steel S-hook is designed for versatile use in marine and industrial environments. Featuring an electrogalvanized finish for enhanced corrosion resistance, this hardware is engineered for reliable performance in various rigging and fastening applications.
  • Opening size of 1/8 in.
  • Load capacity of 7 lb.
  • Durable low-carbon steel construction
  • Electrogalvanized zinc coating for added protection
  • Suitable for both industrial and marine applications