Learn More
MSC Miller Wall Mount Sleeve, Zinc-Plated Steel, 9-1/4 in., 1 ea
Catalog No. 5000508082
Encompass
This wall-mount sleeve is designed for use with approved masts and accessories in fall protection and confined space entry applications. Constructed from welded A-36 steel plate with a zinc chromate finish, it provides reliable structural support for overhead lifting or fall arrest tasks.
  • Enhanced corrosion resistance provided by zinc-plated steel construction
  • Compatible with standard masts for a stable fit and 360-degree rotation
  • Rated for a 450 lbs (204 kg) working load limit with a minimum 4:1 design factor
  • Engineered to withstand a 90,000 in-lbs (10,170 N.m) moment load and 5,000 lb (22.2 kN) vertical load
  • Installation requires approval by a qualified engineer to meet local regulatory standards
Catalog No. 50-005-08082 Supplier MSC Supplier No. 31899982
This wall-mount sleeve is designed for use with approved masts and accessories in fall protection and confined space entry applications. Constructed from welded A-36 steel plate with a zinc chromate finish, it provides reliable structural support for overhead lifting or fall arrest tasks.
  • Enhanced corrosion resistance provided by zinc-plated steel construction
  • Compatible with standard masts for a stable fit and 360-degree rotation
  • Rated for a 450 lbs (204 kg) working load limit with a minimum 4:1 design factor
  • Engineered to withstand a 90,000 in-lbs (10,170 N.m) moment load and 5,000 lb (22.2 kN) vertical load
  • Installation requires approval by a qualified engineer to meet local regulatory standards