Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc Preparation

Catalog No. 499552100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-955-2100ML 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Danger
Catalog No. 49-955-2100ML Supplier Sigma Aldrich Supplier No. 499552100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Rieke™ Zinc metal can be used to reduce common organic functional groups such as aromatic nitro, conjugated aldehydes, arylacetylenes, and phenylpropiolates.Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 7631-86-9
Density 1.23 g/mL at 25°C
Molecular Formula O2Si
Linear Formula SiO2
MDL Number MFCD00011232
Quantity 1 L
Molecular Weight (g/mol) 60.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.