Learn More
Medchemexpress LLC HY-101193 10mg Medchemexpress, Zinc Protoporphyrin CAS:15442-64-5 Purity:>98%
SDP
Catalog No. 501967811
Encompass_Preferred
Medchemexpress, HY-101193 10mg Zinc Protoporphyrin CAS:15442-64-5 Zinc Protoporphyrin (Zn(II)-protoporphyrin IX) is a competitive heme oxygenase-1 (HO-1) inhibitor, markedly attenuates the protective effects of Phloroglucinol (PG) against H2O2. Zinc Protoporphyrin is a normal metabolite formed in trace amounts during heme biosynthesis and used as a screening marker of iron deficiency in individual pregnant women and children, but also to assess population iron status in combination with haemoglobin concentration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7811 Supplier Medchemexpress LLC Supplier No. HY10119310MG
Medchemexpress, HY-101193 10mg Zinc Protoporphyrin CAS:15442-64-5 Zinc Protoporphyrin (Zn(II)-protoporphyrin IX) is a competitive heme oxygenase-1 (HO-1) inhibitor, markedly attenuates the protective effects of Phloroglucinol (PG) against H2O2. Zinc Protoporphyrin is a normal metabolite formed in trace amounts during heme biosynthesis and used as a screening marker of iron deficiency in individual pregnant women and children, but also to assess population iron status in combination with haemoglobin concentration. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.