Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc protoporphyrin | 15442-64-5 | MFCD00011612 | 626.02 | C34H32N4O4Zn | 50 MG
SDP

Catalog No. 5000212044
Encompass_Preferred

Zinc protoporphyrin (Zn(II)-protoporphyrin IX) is a metalloporphyrin research reagent used as a competitive inhibitor of heme oxygenase-1 (HO-1) and as a biomarker in studies of iron deficiency. It is applied in biochemical and cellular assays to probe HO-1 activity and related pathways.

  • Competitive heme oxygenase-1 (HO-1) inhibitor.
  • Used as a biomarker for iron deficiency in vivo.
  • Suitable for biochemical and cellular assays.
  • Molecular formula C34H32N4O4Zn; molecular weight 626.02.
  • Store at -20°C, sealed and protected from moisture; in solvent store at -80°C for long term.

Catalog No. 50-002-12044 Supplier Medchemexpress LLC Supplier No. HY10119350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinc protoporphyrin (Zn(II)-protoporphyrin IX) is a metalloporphyrin research reagent used as a competitive inhibitor of heme oxygenase-1 (HO-1) and as a biomarker in studies of iron deficiency. It is applied in biochemical and cellular assays to probe HO-1 activity and related pathways.

  • Competitive heme oxygenase-1 (HO-1) inhibitor.
  • Used as a biomarker for iron deficiency in vivo.
  • Suitable for biochemical and cellular assays.
  • Molecular formula C34H32N4O4Zn; molecular weight 626.02.
  • Store at -20°C, sealed and protected from moisture; in solvent store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.