Learn More
Cayman Chemical Zinc Protoporphyrin-9, CAS: 15442-64-5, 25 mg
SDP
Catalog No. 502969668
Encompass_Preferred
A competitive inhibitor of heme oxygenase that also inhibits soluble guanylyl cyclase and inactivates the NOS isoforms nNOS, iNOS, and eNOS (IC50s = 0.8, 4, and 5 μM, respectively); used to assess iron status and to diagnose iron disorders and has shown therapeutic potential in hyperbilirubinemia
Catalog No. 50-296-9668 Supplier Cayman Chemical Supplier No. 1448325 MG
A competitive inhibitor of heme oxygenase that also inhibits soluble guanylyl cyclase and inactivates the NOS isoforms nNOS, iNOS, and eNOS (IC50s = 0.8, 4, and 5 μM, respectively); used to assess iron status and to diagnose iron disorders and has shown therapeutic potential in hyperbilirubinemia