Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zinc protoporphyrin (Zn(II)-protoporphyrin IX) | 15442-64-5 | 99.9% | 626.02 g/mol | C34H32N4O4Zn | 1 ML
SDP

Catalog No. 5000211464
Encompass_Preferred

Zinc protoporphyrin (Zn(II)-protoporphyrin IX) is a research reagent that acts as a competitive heme oxygenase-1 (HO-1) inhibitor and a biomarker for iron-deficiency screening. It is supplied as a ready-to-use 10 mM solution in DMSO and as solid preparations for laboratory use. Typical applications include cellular assays, mechanistic studies, and in vivo screening.

  • Ready-to-use 10 mM solution in DMSO.
  • Also available as solid in multiple mass sizes.
  • Competitive heme oxygenase-1 (HO-1) inhibitor for research.
  • Used as a screening marker for iron deficiency in vivo.
  • High purity (approximately 99.9%).
  • Molecular weight 626.02 g/mol; formula C34H32N4O4Zn.
  • Includes product data sheet and SDS for handling information.

Catalog No. 50-002-11464 Supplier Medchemexpress LLC Supplier No. HY10119310MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinc protoporphyrin (Zn(II)-protoporphyrin IX) is a research reagent that acts as a competitive heme oxygenase-1 (HO-1) inhibitor and a biomarker for iron-deficiency screening. It is supplied as a ready-to-use 10 mM solution in DMSO and as solid preparations for laboratory use. Typical applications include cellular assays, mechanistic studies, and in vivo screening.

  • Ready-to-use 10 mM solution in DMSO.
  • Also available as solid in multiple mass sizes.
  • Competitive heme oxygenase-1 (HO-1) inhibitor for research.
  • Used as a screening marker for iron deficiency in vivo.
  • High purity (approximately 99.9%).
  • Molecular weight 626.02 g/mol; formula C34H32N4O4Zn.
  • Includes product data sheet and SDS for handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.