Learn More
Thermo Scientific™ Zinc PVA Fixative Single Vial
Catalog No. R21765 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
12 Vials/Pk.
Catalog No. Quantity
R21765 12 Vials/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R21765 Supplier Thermo Scientific™ Supplier No. R21765
Add to Cart
Add to Cart
This item is not returnable. View return policy
Collect, preserve and permanently stain intestinal parasites with a zinc-based fixative, part of system which comes with industry standard color-coding and sporked caps.
Collect, preserve and permanently stain intestinal parasites with zinc-based Thermo Scientfic™ Zinc PVA Fixative Single Vial, part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
  • Broad range
  • Long shelf life with industry standard color-coded caps and labels
  • Easy to use spork cap for safe collection
  • Convenient packaging, including bulk kits
  • Multilingual patient collection instructions
Note:
Zinc PVA is copper-based.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Zinc PVA Fixative
Description Zinc PVA Fixative Single Vial
Fill Volume 15 mL
Packaging Single vial. Packaged in partitioned boxes.
Quantity 12 Vials/Pk.