Learn More
Cayman Chemical Zinc Pyrithione, Purity: ≥95%, CAS: 13463-41-7, 100 mg
SDP
Catalog No. 502939970
Encompass_Preferred
A coordination complex of zinc and pyrithione that has antimicrobial and anticancer activities; active against various bacteria (MICs = 1-4 µg/ml) and the fungus P. ovale at 0.01-10 µg/ml; reduces tumor growth in an SCC-4 mouse xenograft model at 1 mg/kg per week for six weeks; topical application completely prevents Aspergillus growth in soy meal-derived adhesives
Catalog No. 50-293-9970 Supplier Cayman Chemical Supplier No. 29154100 MG
A coordination complex of zinc and pyrithione that has antimicrobial and anticancer activities; active against various bacteria (MICs = 1-4 µg/ml) and the fungus P. ovale at 0.01-10 µg/ml; reduces tumor growth in an SCC-4 mouse xenograft model at 1 mg/kg per week for six weeks; topical application completely prevents Aspergillus growth in soy meal-derived adhesives