Learn More
Selleck Chemical LLC Zinc Pyrithione S4075-50mg
SDP
Supplier:  Selleck Chemical LLC S407550MG
Encompass_Preferred
Zinc pyrithione (OM-1563) is an antifungal and antibacterial agent disrupting membrane transport by blocking the proton pump
Catalog No. 50-310-9653