Learn More
Medchemexpress LLC Zinc pyrithione | 13463-41-7 | 98.2% | 5 G
SDP
Catalog No. 5000407913
Encompass_Preferred
Zinc Pyrithione is an antifungal and antibacterial agent that disrupts membrane transport by blocking the proton pump. It is also a copper ionophore that delivers copper into cells and serves as a useful tool for studying cuproptosis.
  • Disrupts membrane transport by blocking the proton pump
  • Serves as a copper ionophore to deliver copper into cells
  • Used for studying cuproptosis
  • Induces plasma membrane depolarization
  • Significantly induces cell death in AAVS1 cells
Catalog No. 50-004-07913 Supplier Medchemexpress LLC Supplier No. HYB05725G
Add to Cart
Add to Cart
Zinc Pyrithione is an antifungal and antibacterial agent that disrupts membrane transport by blocking the proton pump. It is also a copper ionophore that delivers copper into cells and serves as a useful tool for studying cuproptosis.
  • Disrupts membrane transport by blocking the proton pump
  • Serves as a copper ionophore to deliver copper into cells
  • Used for studying cuproptosis
  • Induces plasma membrane depolarization
  • Significantly induces cell death in AAVS1 cells