Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Zinc, Sheet
SCI_ED

Catalog No. S25638A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
6 x 12 in.
12 x 12 in.
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25638A 6 x 12 in. Solid
S25638 12 x 12 in. Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25638A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reacts with water or moisture

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 6-6-7440

Specifications

Color Gray
Density 7.14g/mL
Quantity 6 x 12 in.
Packaging Solid
Assay Percent Range 100% w/w
Linear Formula Zn
Synonym Zinc Dust
Formula Weight 65.38
Chemical Name or Material Zinc, Sheet
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.