Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Zinc, Shot
SCI_ED

Catalog No. S25639 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Reagent Grade
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25639 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S25639
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reacts with water or moisture

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 6-6-7440
Synonym Zinc Dust

Specifications

Density 7.14g/mL
Quantity 100 g
Packaging Reagent Grade
Assay Percent Range 100% w/w
Linear Formula Zn
Formula Weight 65.38
Chemical Name or Material Zinc, Shot
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.